Learn More
Honeywell™ Tuff-Coat™ Cut-Resistant Gloves
Seamless knit liners with durable quality coatings
Manufacturer:  Honeywell™ KV200XXL
 View more versions of this product
Catalog No. 19-175-453
Add to cart
Description
Description
Ideal for construction, forestry, municipal services, and transportation.
  • Standard weight, 7-gauge Kevlar™ seamless knit shell
  • Kevlar provides outstanding cut and slash protection
  • Natural rubber palm and fingertip coating
  • Rubber coating provides excellent grip and flexibility
  • This product contains natural rubber
Compliance
Meets ANSI A2 (cut level) standards.
Specifications
Specifications
Knit
construction, forestry, municipal services and transportation
2X-Large
Blue, Yellow
Orange
Dupont™ Kevlar™ Aramid Fiber
Medium weight