Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Tuff-Coat™ Cut-Resistant Gloves

Seamless knit liners with durable quality coatings

$16.00

Specifications

Coating Material Rough
Color Blue,Yellow
Cut Resistance Rating Level 2
Material Dupont™ Kevlar™ Aramid Fiber
For Use With (Application) construction, forestry, municipal services and transportation
View More Specs

Products 5
Catalog Number Mfr. No. Size Hem Color Price Quantity & Availability  
Catalog Number Mfr. No. Size Hem Color Price Quantity & Availability  
19-175-449
View Documents
Honeywell™
KV200S
Small Red
Pair for $16.00
Case of 144 PR for $2,304.00
Add to cart
 
19-175-451
View Documents
Honeywell™
KV200L
Large Yellow
Pair for $16.00
Case of 144 PR for $2,304.00
Add to cart
 
19-175-450
View Documents
Honeywell™
KV200M
Medium Blue
Pair for $16.00
Case of 144 PR for $2,304.00
Add to cart
 
19-175-452
View Documents
Honeywell™
KV200XL
X-Large White
Pair for $16.00
Case of 144 PR for $2,304.00
Add to cart
 
19-175-453
View Documents
Honeywell™
KV200XXL
2X-Large Orange
Pair for $16.00
Case of 144 PR for $2,304.00
Add to cart
 
Description

Description

Ideal for construction, forestry, municipal services, and transportation.

  • Standard weight, 7-gauge Kevlar™ seamless knit shell
  • Kevlar provides outstanding cut and slash protection
  • Natural rubber palm and fingertip coating
  • Rubber coating provides excellent grip and flexibility
  • This product contains natural rubber

Compliance

Meets ANSI A2 (cut level) standards.

Specifications

Specifications

Rough
Level 2
construction, forestry, municipal services and transportation
Medium weight
Blue,Yellow
Dupont™ Kevlar™ Aramid Fiber
Knit
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit