Honeywell Tuff-Coat™ Cut-Resistant Gloves

Seamless knit liners with durable quality coatings

Manufacturer: Honeywell KV200XL

 View more versions of this product

Catalog No. 19-175-452

  Check Availability
Add to Cart

    Specifications

    Glove Cuff Style Knit
    Hem Color White
    For Use With (Application) construction, forestry, municipal services and transportation
    Material Kevlar™ Fabric
    Size X-Large
    Weight Medium weight
    Coating Material Rough
    Color Yellow, blue
    Cut Resistance Rating Level 2

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit