Learn More
Honeywell™ Tuff-Coat™ Dipped Gloves
Feature seamless knit liners and durable quality coatings
Manufacturer:  Honeywell™ 125S
 View more versions of this product
Catalog No. 19-175-435
Add to cart
Description
Description
  • Available in a variety of fibers to suit various application needs
  • Soft and comfortable, with no seams to rub and chafe
  • Coatings available in natural rubber and nitrile in many configurations
  • Coatings provide extra protection and help to inhibit liquid penetration
Specifications
Specifications
Palm, finger
Rough
Provides extra protection, inhibit liquid penetration
Nylon
Blue, White
12 in.
Withstands commercial laundry room detergents (up to pH 14) and warm water temperatures up to 120°F. Lower heat settings up to 180°F are suggested for drying as extreme heat may cause shrinking and degradation.
Elastic
Latex
Small
Red
30.5 cm