Learn More
Honeywell™ Tuff-Coat II™ Cut-Resistant Gloves
Manufacturer:  Honeywell™ KV300S
Catalog No. 19-050-003A
Add to cart
Specifications
Specifications
Safety
Level 2
Red
Dupont™ Kevlar™ Aramid Fiber
Medium weight
Rough
Knit
handling Glass, small parts and bricks, canning, recycling, laundering, integrated waste management, general maintenance, construction, light fabrication, sanitation, farming, lumbering, mill work and shipping and receiving
Small
Blue, Yellow