Learn More
Honeywell™ Tuff-Knit KV Lightweight Gloves
For the ultimate in performance, comfort and fit
Manufacturer:  Honeywell™ KVSP13A
Catalog No. 19-050-337
Add to cart
Description
Description
  • Kevlar™/Lycra™ blend fiber construction offers exceptional cut, slash, and heat resistance
  • Seamless construction provides superior fit and comfort with less hand fatigue
  • Lightweight yet durable
  • Launderable
  • Men's size
Specifications
Specifications
Safety
automotive and part manufacturer, building and construction, fishing, Glass industries, industrial cleaning, maintenance, Metal Steel, Paper industries, ship building
Men's
Yellow
Knit
Dupont™ Kevlar™ Aramid Fiber, Lycra™
Light weight