Learn More
Honeywell™ Unisex Knee Hi Safety Boots
Comfortable olive-colored work boots ideal for general use, farms and occupational applications that provide excellent protection
Manufacturer:  Honeywell™ 75120ODM120
 View more versions of this product
Catalog No. 19-120-660
Add to cart
Description
Description
  • 100% waterproof
  • TDT (Triple Density Technology) uses proprietary compound formulations for increased resistance to degradation from many harsh chemicals
  • Outsole made of polyvinyl chloride material
  • Centersole is dual compounded for cushioning, long wear and traction on slick surfaces
  • Foot Form contour insole provides additional comfort
  • Reinforced construction at critical stress points provides extended durability
  • Higher, wider, kick-off lufs offer easy on/off convenience
Order Info
Must be ordered in men's sizes, unless there are specified women's sizes.