Honeywell Unisex Knee Hi Safety Boots
Comfortable olive-colored work boots ideal for general use, farms and occupational applications that provide excellent protection
Manufacturer: Honeywell 75120ODM030
 View more versions of this product
Catalog No. 19-120-651
  Check Availability
Add to Cart
    Specifications
    Size 3P
    Product Type Knee High Safety Boots
    Material Poly vinyl
    Height (English) 16 in.
    Height (Metric) 40.6cm
    Toe Material Steel
    Color Olive green
  • Description & Specifications
Related Products
Product Content Feedback