Learn More
Honeywell™ V-Safe Wheel Valve Lockouts
Manufacturer:  Honeywell™ VS02G
 View more versions of this product
Catalog No. 19-170-162
Add to cart
Specifications
Specifications
Wheel Valve Lockout
2.75 in.
7cm
Manufacturing, mining, municipal services, oil and gas, utilities
3.2cm
0.31 in.
-50° to 360°F
7 lb.
Polypropylene
Green
3 in.
8cm
1.25 in.
Two flattened half moons
0.8cm
-46° to 182°C
3kg