Honeywell Ventilated Eagle Hoods
Double-layer head and bib construction with elastic-reinforced face opening
Manufacturer: Total Fire Group HDFCP84
 View more versions of this product
Catalog No. 19-815-032
  Check Availability
Add to Cart
    Specifications
    Color Navy
    Material P-84/PFR Rayon
    • Reduce firefighter heat stress by trapping less heat than traditional hood styles
    • Pbi™ open-weave mesh ventilating panel in crown allows maximum heat release from top of head
    • Sewn with 100% Nomex™ thread
    Available in two styles:
  • Ventilated Face and Chest features Pbi mesh ventilating panel with double neck and full 3.5 in. (8.9cm) chest/back drapes that provide extra interface overlap at the chest and back
  • Ventilated Full Drape features Pbi mesh ventilating panel with double layer head/neck and full 8 in. (20.3cm) chest/back drapes that provide extra interface overlap at the chest and back
  • Description & Specifications
  • Related Products
    Product Content Feedback