Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Whiting & Davis™ Metal Mesh Gloves
Click to view available options
Size:
X-Large
Description
- Individually welded rings allow the mesh fabric to flow without harsh edges
- Seams are 4-ring joined, sealing the seam so gaps are non-existent
- Thumbs are attached with a 3-ring joint for a strong seam at a high wear area
- Ergonomically designed and constructed with tapered fingers and palms to prevent sagging
- Gloves are 3-step reversible to be worn on either hand
- Mesh is manufactured following the European Standard of EN1082
Specifications
Color | Silver |
Hand | Ambidextrous |
Material | Stainless Steel Fiber |
Product Type | Cut-resistant Gloves |
Size | X-Large |
For Use With (Application) | food service, knife and blade sharpening/cleaning, fishing, garment manufacturing, veterinary services, animal control, Paper and pulp industry |
Glove Cuff Style | Strap with dome fastener |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction