Learn More
Honeywell™ WorkEasy™ General Purpose Gloves
Nitrile coating helps to reduce nicks, cuts, and abrasions
Manufacturer:  Honeywell™ WE110XL
 View more versions of this product
Catalog No. 19-175-473
Add to cart
Description
Description
Polyester liners are seamless and lightweight for excellent dexterity and tactile sensitivity.
  • Palm dipped design helps reduce hand fatigue and offers excellent dexterity
  • Liners keep hands cool and protected from dirt, incidental cuts and abrasions
  • Excellent tactile sensitivity
  • Variety of color combinations
Specifications
Specifications
Palm, finger
Knit
Nitrile
X-Large
Smooth
excellent dexterity and tactile sensitivity
Polyester
Black, White