Learn More
Honeywell WorkEasy Polyurethane Coated Gloves
Excellent for work in construction or municipal services
Manufacturer: Honeywell WE200S
Description
- Lightweight 13-gauge liners keep hands cool and protected from dirt, incidental cuts and abrasions
- Thin polyurethane coating offers added protection and grip
- Excellent tactile sensitivity
- Palm dipped design helps reduce hand fatigue and offers excellent dexterity
- Polyester liners are seamless and lightweight for excellent dexterity and sensitivity
- Polyurethane coating is flexible, and excellent for grip
- HPPE (High Performance Polyethylene) fiber liner for added cut protection
- Durable polyurethane coating for added grip and durability
- ANSI cut level 3; EN388 cut level 5
Specifications
Palm, finger | |
Knit | |
Polyurethane | |
Small | |
Red |
Smooth | |
work in construction or municipal services | |
Polyester | |
Gray, Red, White |