Learn More
Honeywell™ WorkEasy™ Polyurethane Coated Gloves
Excellent for work in construction or municipal services
Manufacturer:  Honeywell™ WE210M
 View more versions of this product
Catalog No. 19-175-481
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Lightweight 13-gauge liners keep hands cool and protected from dirt, incidental cuts and abrasions
  • Thin polyurethane coating offers added protection and grip
  • Excellent tactile sensitivity
  • Palm dipped design helps reduce hand fatigue and offers excellent dexterity
WE200 and WE210
  • Polyester liners are seamless and lightweight for excellent dexterity and sensitivity
  • Polyurethane coating is flexible, and excellent for grip
WE300
  • HPPE (High Performance Polyethylene) fiber liner for added cut protection
  • Durable polyurethane coating for added grip and durability
  • ANSI™ cut level 3; EN388 cut level 5
Specifications
Specifications
Palm, finger
Knit
Polyurethane
Medium
Blue
Smooth
work in construction or municipal services
Polyester
Black, Blue, Gray