Honeywell WorkEasy™ Polyurethane Coated Gloves

Excellent for work in construction or municipal services

Manufacturer: Honeywell WE200XL

 View more versions of this product

Catalog No. 19-175-478

  Check Availability
Add to Cart

    Specifications

    Exterior Finish Smooth
    Glove Cuff Style Knit
    Glove Cuff Color White
    For Use With (Application) work in construction or municipal services
    Material Polyurethane
    Liner Material Polyester
    Size X-Large
    Coated Area Palm, finger
    Color Gray, white
    Cut Resistance Rating Level 3

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit