Honeywell WorkEasy™ Polyurethane Coated Gloves

Excellent for work in construction or municipal services

Manufacturer: Honeywell WE210S

 View more versions of this product

Catalog No. 19-175-480

  Check Availability
Add to Cart

    Specifications

    Exterior Finish Smooth
    Glove Cuff Style Knit
    For Use With (Application) work in construction or municipal services
    Liner Material Polyester
    Cut Resistance Rating Level 3
    Glove Cuff Color Red
    Material Polyurethane
    Size Small
    Coated Area Palm, finger
    Color Black, gray, red

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit