Learn More
Medicom™ Hopen™ Anti-Static Hygiene Gown
Medicom™ Hopen™ Anti-Static Hygiene Gown with a waterproof, microporous polyethylene lining
$218.82
Specifications
Certifications/Compliance Manufactured under an ISO 9001 Certified Quality System
Certifications/Compliance (Europe) EN 1149-5
Closure Type Zipper
Color White
Construction Made in China
View More Specs
Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-910-577
View Documents
Medicom™
40.507/03
Large
Case of 50 for $218.82
Add to cart
19-910-578
View Documents
Medicom™
40.507/04
X-Large
Case of 50 for $218.82
Add to cart
19-910-579
View Documents
Medicom™
40.507/05
2X-Large
Case of 50 for $218.82
Add to cart
19-910-580
View Documents
Medicom™
40.507/06
3X-Large
Case of 50 for $218.82
Add to cart
Description
Description
  • Single use, anti-static hygiene gown provides the user with full body protection
  • Protects against activity related soiling and contamination
  • Microporous, polyethylene lining is comfortable, and waterproof
Specifications
Specifications
Manufactured under an ISO 9001 Certified Quality System
Zipper
Made in China
Yes
Unisex
None
Hygiene Gown
Non-sterile
EN 1149-5
White
Elastic
Category 1 Personal Protective Equipment
Polypropylene, SMS, Microporous Polyethylene Laminate
Individual, Bag
Sewn