Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Medicom™ Hopen™ Double Elastic Bouffant cap
Click to view available options
Color:
Pink
White
Yellow
Description
- Lightweight, polypropylene bouffant cap provides additional comfort the user
- Full head coverage ensures excellent retention for the user
- Ability to use a color code for tracking
- Can be used to control the environment for HACCP processes and various industries
Specifications
Disposable | Single-use |
For Use With (Equipment) | Operating Room PPE |
Width (Metric) | 50 cm |
Construction | Made in China |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction