Learn More
Medicom™ Hopen™ HOP'CLEAN Hygiene Coveralls
Medicom™ Hopen™ HOP'CLEAN Hygiene Coveralls in SMS-Polypropylene limits particulate release (lint-free)
$176.27
Specifications
Ankle Style Elastic
Certifications/Compliance Manufactured under an ISO 9001 Certified Quality System
Color Blue
Cuff Style Thumb loops
Disposable Yes
View More Specs
Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-910-608
View Documents
Medicom™
40.036C/03
Large
Case of 25 for $176.27
Add to cart
19-910-609
View Documents
Medicom™
40.036C/04
X-Large
Case of 25 for $176.27
Add to cart
19-910-610
View Documents
Medicom™
40.036C/05
2X-Large
Case of 25 for $176.27
Add to cart
19-910-611
View Documents
Medicom™
40.036C/06
3X-Large
Case of 25 for $176.27
Add to cart
Description
Description
  • HOP'CLEAN Hygiene Coveralls protect the user from activity related soiling
  • Pleasant and light, SMS Polypropylene material is breathable but offers a better seal than a simple polypropylene
  • Suit offers good resistance to tearing and abrasion
  • Permeable to air and water vapors
Specifications
Specifications
Elastic
Blue
Yes
Unisex
Collared
Coveralls
Manufactured under an ISO 9001 Certified Quality System
Thumb loops
Category 1 Personal Protective Equipment
Polypropylene, SMS
None
Non-sterile