Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Hopen™ HOP'SAFE EXTRA Coveralls

Click to view available options
Size:
Small
Medium
Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Medicom™ Hopen™ HOP'SAFE EXTRA Coveralls protect against radioactive particles, and infectious agents

  • These HOP'SAFE EXTRA coveralls are hooded with an adhesive flap on the zipper closure
  • Antistatic treatment, radioactive particle protection, infectious agent protection
  • Waterproof Polypropylene SMS with Polyethylene membrane is air and water vapor permeable
  • Coverall EPI, Category III, Type 4B, 5B, 6B
  • Elasticated wrists, ankles, hood, and waist
  • Lint-free material is tear and abrasion resistant
  • Single use, non sterile
  • Individually packaged
Product Type Multiple Use Coveralls
Size Large
Gender Unisex
Material Polypropylene, Polyethylene
Color White
Disposable No
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper
Sterility Non-sterile
Resistance Anti-static, Particle
Certifications/Compliance (Europe) EN 1149-5, EN 1073-2, EN 14126
No. of Pockets None
Thickness 65 g/m2
For Use With (Application) Category III, 4B/5B/6B PPE
Show More Show Less