Learn More
Medicom™ Hopen™ HOP'SAFE EXTRA Coveralls
Medicom™ Hopen™ HOP'SAFE EXTRA Coveralls protect against radioactive particles, and infectious agents
$628.00 - $693.00
Specifications
Ankle Style Elastic
Certifications/Compliance (Europe) EN 1149-5, EN 1073-2, EN 14126
Closure Type Zipper
Color White
Cuff Style Elastic
View More Specs
Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-910-514
View Documents
Medicom™
40.070/01
Small
Case of 25 for $628.00
Add to cart
19-910-513
View Documents
Medicom™
40.070/02
Medium
Case of 25 for $693.00
Add to cart
19-910-512
View Documents
Medicom™
40.070/04
Large
Case of 25 for $693.00
Add to cart
19-910-515
View Documents
Medicom™
40.070/05
2X-Large
Case of 25 for $693.00
Add to cart
19-910-516
View Documents
Medicom™
40.070/06
3X-Large
Case of 25 for $693.00
Add to cart
Description
Description
  • These HOP'SAFE EXTRA coveralls are hooded with an adhesive flap on the zipper closure
  • Antistatic treatment, radioactive particle protection, infectious agent protection
  • Waterproof Polypropylene SMS with Polyethylene membrane is air and water vapor permeable
  • Coverall EPI, Category III, Type 4B, 5B, 6B
  • Elasticated wrists, ankles, hood, and waist
  • Lint-free material is tear and abrasion resistant
  • Single use, non sterile
  • Individually packaged
Specifications
Specifications
Elastic
Zipper
Elastic
Category III, 4B/5B/6B PPE
Polypropylene, Polyethylene
None
Anti-static, Particle
65 g/m2
EN 1149-5, EN 1073-2, EN 14126
White
No
Unisex
Hooded
Multiple Use Coveralls
Non-sterile
Elastic