Learn More
Medicom™ Hopen™ HOP'SAFE Hooded Coveralls
Medicom™ Hopen™ HOP'SAFE Hooded Coveralls are anti-static, and protect against radioactive particles
$345.51
Specifications
Ankle Style Elastic
Certifications/Compliance (Europe) EN1149-5, EN1073-2
Closure Type Zipper
Color White
Cuff Style Elastic
View More Specs
Products 6
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-910-585
View Documents
Medicom™
40.040/01
Small
Case of 25 for $345.51
Add to cart
19-910-584
View Documents
Medicom™
40.040/02
Medium
Case of 25 for $345.51
Add to cart
19-910-583
View Documents
Medicom™
40.040/03
Large
Case of 25 for $345.51
Add to cart
19-910-586
View Documents
Medicom™
40.040/04
X-Large
Case of 25 for $345.51
Add to cart
19-910-587
View Documents
Medicom™
40.040/05
2X-Large
Case of 25 for $345.51
Add to cart
19-910-588
View Documents
Medicom™
40.040/06
3X-Large
Case of 25 for $345.51
Add to cart
Description
Description
  • These HOP'SAFE coveralls are hooded with an adhesive flap on the zipper closure
  • Polypropylene SMS material is air and water vapor permeable
  • Coverall EPI, Category III, Type 5/6
  • External seams provide additional comfort
  • Good tear, and abrasion resistance
  • Single use, non sterile
  • Individually packaged
Specifications
Specifications
Elastic
Zipper
Elastic
Category III, Type 5/6 PPE
Hooded
None
Anti-static, Particle
55 g/m2
EN1149-5, EN1073-2
White
Yes
Unisex
Polypropylene, SMS
Single Use Coveralls
Non-sterile
Elastic