Learn More
Medicom™ Hopen™ HOP'SAFE PLUS Coveralls
Medicom™ Hopen™ HOP'SAFE PLUS Coveralls with a microporous membrane can be used multiple times
$628.00
Specifications
Ankle Style Elastic
Certifications/Compliance (Europe) EN 1149-5, EN 1073-2, EN 14126
Closure Type Zipper
Color White
Cuff Style Elastic
View More Specs
Products 12
Catalog Number Mfr. No. Thickness Size Price Quantity & Availability  
Catalog Number Mfr. No. Thickness Size Price Quantity & Availability  
19-910-502
View Documents
Medicom™
40.050/01
60 g/m2 Small
Case of 25 for $628.00
Add to cart
19-910-501
View Documents
Medicom™
40.050/02
60 g/m2 Medium
Case of 25 for $628.00
Add to cart
19-910-500
View Documents
Medicom™
40.050/03
60 g/m2 Large
Case of 25 for $628.00
Add to cart
19-910-503
View Documents
Medicom™
40.050/04
60 g/m2 X-Large
Case of 25 for $628.00
Add to cart
19-910-504
View Documents
Medicom™
40.050/05
60 g/m2 2X-Large
Case of 25 for $628.00
Add to cart
19-910-505
View Documents
Medicom™
40.050/06
60 g/m2 3X-Large
Case of 25 for $628.00
Add to cart
19-910-508
View Documents
Medicom™
40.055/01
65 g/m2 Small
Case of 25 for $628.00
Add to cart
19-910-507
View Documents
Medicom™
40.055/02
65 g/m2 Medium
Case of 25 for $628.00
Add to cart
19-910-506
View Documents
Medicom™
40.055/03
65 g/m2 Large
Case of 25 for $628.00
Add to cart
19-910-509
View Documents
Medicom™
40.055/04
65 g/m2 X-Large
Case of 25 for $628.00
Add to cart
19-910-510
View Documents
Medicom™
40.055/05
65 g/m2 2X-Large
Case of 25 for $628.00
Add to cart
19-910-511
View Documents
Medicom™
40.055/06
65 g/m2 3X-Large
Case of 25 for $628.00
Add to cart
Description
Description
  • These HOP'SAFE PLUS coveralls are hooded with an adhesive flap on the zipper closure
  • Antistatic treatment, radioactive particle protection, infectious agent protection
  • Microporous membrane is permeable to air and water vapor
  • Polypropylene SMS Enduction PE
  • Coverall EPI, Category III, Type 5B/6B
  • Elasticated wrists, ankles, hood, and waist
  • Lint-free material is tear and abrasion resistant
  • Can be used multiple times
  • Taped, internal seams
  • Individually packaged
Specifications
Specifications
Elastic
Zipper
Elastic
Category III, Type 5B/6B PPE
Polypropylene, SMS
None
Anti-static, Particle
Non-sterile
EN 1149-5, EN 1073-2, EN 14126
White
No
Unisex
Hooded
Multiple Use Coveralls
Taped
Elastic