Learn More
Medicom™ Hopen™ HOP'SAFE PRO Coveralls
Medicom™ Hopen™ HOP'SAFE PRO Coveralls are extremely resistant and offer users maximum protection
$345.51
Specifications
Ankle Style Elastic
Color Blue
Cuff Style Elastic
Disposable Yes
Gender Unisex
View More Specs
Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-910-589
View Documents
Medicom™
40.035/02
Medium
Case of 25 for $345.51
Add to cart
19-910-590
View Documents
Medicom™
40.035/03
Large
Case of 25 for $345.51
Add to cart
19-910-591
View Documents
Medicom™
40.035/04
X-Large
Case of 25 for $345.51
Add to cart
19-910-592
View Documents
Medicom™
40.035/05
2X-Large
Case of 25 for $345.51
Add to cart
19-910-593
View Documents
Medicom™
40.035/06
3X-Large
Case of 25 for $345.51
Add to cart
Description
Description
  • HOP'SAFE PRO Coveralls offer Category I protection
  • Lint-free material is tear and abrasion resistant
  • Offer users maximum protection
Specifications
Specifications
Elastic
Elastic
Unisex
Polypropylene, SMS
None
Non-sterile
Blue
Yes
Elastic
Hooded
Single Use Coveralls
50 g/m2