Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Hopen™ Hygiene Eco Coveralls

Catalog Number 19910618
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Medicom™ Hopen™ Hygiene Eco Coveralls are suitable to HACCP procedures

  • These Hygiene Eco white coveralls protect the working environment from particles coming from the body of the operator
  • Allows the operator to protect himself from the risks of soiling related to the activity being carried out
  • Category 1 Personal Protective Equipment
  • Suitable to HACCP procedures
Product Type Single-use Coveralls
Size Large
Gender Unisex
Material Polypropylene
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Closure Type Zipper
Seam Type Stitched
Sterility Non-sterile
No. of Pockets None
For Use With (Application) Category 1 Personal Protective Equipment
Weight (Metric) 30 g/m2
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit