Learn More
Medicom™ Hopen™ Hygiene Eco Coveralls
Medicom™ Hopen™ Hygiene Eco Coveralls are suitable to HACCP procedures
$218.82
Specifications
Ankle Style Elastic
Closure Type Zipper
Color White
Cuff Style Elastic
Disposable Yes
View More Specs
Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-910-617
View Documents
Medicom™
40.103/02
Medium
Case of 25 for $218.82
Add to cart
19-910-618
View Documents
Medicom™
40.103/03
Large
Case of 25 for $218.82
Add to cart
19-910-619
View Documents
Medicom™
40.103/04
X-Large
Case of 25 for $218.82
Add to cart
19-910-620
View Documents
Medicom™
40.103/05
2X-Large
Case of 25 for $218.82
Add to cart
Description
Description
  • These Hygiene Eco white coveralls protect the working environment from particles coming from the body of the operator
  • Allows the operator to protect himself from the risks of soiling related to the activity being carried out
  • Category 1 Personal Protective Equipment
  • Suitable to HACCP procedures
Specifications
Specifications
Elastic
White
Yes
Unisex
Collared
Single Use Coveralls
Non-sterile
Zipper
Elastic
Category 1 Personal Protective Equipment
Polypropylene
None
Stitched
30 g/m2