Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Hopen™ Single Use Hygiene Gown

Catalog Number 19910635
Click to view available options
Closure Type:
Press Studs
Zipper
Size:
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Medicom™ Hopen™ Single Use Hygiene Gown without pockets, is microporous and waterproof

  • Reduces contamination of the operator, but also sensitive products and processes in a protected atmosphere
  • Protects against radioactive particles and infectious agents
  • Microporous, Anti-Static Hygiene Gown is comfortable and light
  • Polypropylene gown lined with laminated polyethylene for a waterproof, hydrophobic seal
  • Available in mulitple sizes, and closure types
Size Large
Product Type General Purpose Gown
Certifications/Compliance (Europe) EN 1149-5
Construction Made in China
For Use With (Application) Category 1 Personal Protective Equipment
Gender Unisex
No. of Pockets None
Packaging Type Individual, Bag
Material Polypropylene
Color White
Closure Type Press Studs
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Finish Microporous, Waterproof
Seam Type Sewn
Resistance Radioactive Particles, Infectious Agents
Certifications/Compliance ISO 9001
Autoclavable Not Autoclavable
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit