Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Hopen™ Pocketed, Multiple Use Hygiene Gown

Click to view available options
Color:
Blue
White
Closure Type:
Press studs
Zipper
Size:
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Medicom™ Hopen™ Pocketed, Multiple Use Hygiene Gown available in multiple colors

  • Single use hygiene gowns protect the working environment from particles coming from the body of the operator (dander, hair, sweating ...)
  • They are suitable to HACCP procedures.
  • They allow the operator to protect himself from the risks of soiling related to the activity he carries out.
Type Hygiene Gown
Size X-Large
Gender Unisex
Material Polypropylene
Construction Made in China
Color White
Closure Type Press studs
Cuff Style Elastic
Disposable Yes
Sterility Non-sterile
No. of Pockets 2
Seam Type Sewn
Certifications/Compliance Manufactured under an ISO 9001 Certified Quality System
For Use With (Application) Category 1 Personal Protective Equipment
Packaging Type Individual, Bag
Show More Show Less