Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Hopen™ Pocketless Hygiene Gown

Catalog Number 19910560
Click to view available options
Color:
Green
Red
White
Size:
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Medicom™ Hopen™ Pocketless Hygiene Gown available in three distinct colors

  • The Pocketless Hygiene gown protects the working environment from particles coming from the body of the operator
  • Allows the user to be protected from the risks of soiling related to the activity being carried out
  • Pleasant and light, polypropylene fabric is extremely breathable
  • Collared model without flap and elasticated at the wrists
Type Hygiene Gown
Size X-Large
Gender Unisex
Material Polypropylene
Color White
Closure Type Press Studs
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
No. of Pockets None
Seam Type Sewn
Certifications/Compliance Manufactured under an ISO 9001 Certified Quality System
For Use With (Application) Category 1 Personal Protective Equipment
Packaging Type Individual, Bag
Construction Made in China
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit