Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Hopen™ Polypropylene Coveralls

Catalog Number 19910603
Click to view available options
Color:
Blue
White
Neck Style:
Collared
Hooded
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Medicom™ Hopen™ Polypropylene Coveralls available in collared, or hooded styles

  • Hygiene Classic Polypropylene Coveralls protect the working environment from particles coming from the body of the operator
  • Protects the operator from the risks of soiling related to their purpose for use
  • Polypropylene fabric is light, and breathable
  • Elasticated at cuffs, waist, and ankles
  • Individually packaged for controlled hygiene and distribution
  • Category 1 Personal Protective Equipment
Product Type Cleanroom Coveralls
Size X-Large
Gender Unisex
Material ComforTech™ Fabric
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper
Seam Type Stitched
Sterility Non-sterile
Certifications/Compliance Manufactured under an ISO 9001 Certified Quality System
No. of Pockets None
For Use With (Application) Controlled Environments
Weight (Metric) 40 g/m2
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit