Learn More
Medicom™ Hopen™ Polypropylene Coveralls
Medicom™ Hopen™ Polypropylene Coveralls available in collared, or hooded styles
$240.00
Specifications
Ankle Style Elastic
Certifications/Compliance Manufactured under an ISO 9001 Certified Quality System
Closure Type Zipper
Cuff Style Elastic
Disposable Yes
View More Specs
Products 11
Catalog Number Mfr. No. Color Neck Style Size Price Quantity & Availability  
Catalog Number Mfr. No. Color Neck Style Size Price Quantity & Availability  
19-910-621
View Documents
Medicom™
40.205BM/03
Blue Collared Large
Case of 25 for $240.00
Add to cart
19-910-622
View Documents
Medicom™
40.205BM/04
Blue Collared X-Large
Case of 25 for $240.00
Add to cart
19-910-623
View Documents
Medicom™
40.205BM/05
Blue Collared 2X-Large
Case of 25 for $240.00
Add to cart
19-910-624
View Documents
Medicom™
40.205BM/06
Blue Collared 3X-Large
Case of 25 for $240.00
Add to cart
19-910-604
View Documents
Medicom™
40.201BM/02
Blue Hooded Medium
Case of 25 for $240.00
Add to cart
19-910-605
View Documents
Medicom™
40.201BM/03
Blue Hooded Large
Case of 25 for $240.00
Add to cart
19-910-606
View Documents
Medicom™
40.201BM/04
Blue Hooded X-Large
Case of 25 for $240.00
Add to cart
19-910-607
View Documents
Medicom™
40.201BM/05
Blue Hooded 2X-Large
Case of 25 for $240.00
Add to cart
19-910-601
View Documents
Medicom™
40.201/01
White Hooded Small
Case of 25 for $240.00
Add to cart
19-910-602
View Documents
Medicom™
40.201/03
White Hooded Large
Case of 25 for $240.00
Add to cart
19-910-603
View Documents
Medicom™
40.201/04
White Hooded X-Large
Case of 25 for $240.00
Add to cart
Description
Description
  • Hygiene Classic Polypropylene Coveralls protect the working environment from particles coming from the body of the operator
  • Protects the operator from the risks of soiling related to their purpose for use
  • Polypropylene fabric is light, and breathable
  • Elasticated at cuffs, waist, and ankles
  • Individually packaged for controlled hygiene and distribution
  • Category 1 Personal Protective Equipment
Specifications
Specifications
Elastic
Zipper
Yes
Unisex
Polypropylene
Stitched
Elasticated
40 g/m2
Manufactured under an ISO 9001 Certified Quality System
Elastic
Category 1 Personal Protective Equipment
No
Single Use Coveralls
Non-sterile
None