Learn More
Medicom™ Hopen™ Type IIR Folding Face Masks
Folding masks protect users against various industrial hazards stipulated in N95 standards.
Supplier: Medicom™ M72010SWH
This item is not returnable.
View return policy
Description
- Filtering half mask, efficency class FFP2 (D)
- Protects users against moderate levels of fine dust particles, and oil or water based mist
- Hazards are initially identified and evaluated by the employer as stipulated in N95 standards
- Typical applications include plastering, cement, sanding, and wood dust
- Prevents against risky infectious diseases, according to national recommendations
- Typical use time: 8 hours
Specifications
General Purpose Face Mask | |
Ties | |
Non-sterile | |
Yes |
White | |
Polypropylene | |
EN 14683 | |
M72010S-WH |