Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Hopen™ Type IIR Folding Mask

Medicom™ Hopen™ Type IIR Folding Mask protects users against various industrial hazards stipulated in N95 standards

Specifications

Certifications/Compliance (Europe) EN 14683 Type: IIR
Construction Made in France
Dimensions (L x W x H) 240 x 90 mm (± 3 mm)
Disposable Yes
Filter N95
View More Specs

Products 3
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
19-910-518
View Documents
Medicom™
M72010SBL
Blue
Case of 10 for $351.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-910-519
View Documents
Medicom™
M72010SGR
Green
Case of 10 for $351.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-910-517
View Documents
Medicom™
M72010SWH
White
Case of 10 for $351.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

  • Filtering half mask, efficency class FFP2 (D)
  • Protects users against moderate levels of fine dust particles, and oil or water based mist
  • Hazards are initially identified and evaluated by the employer as stipulated in N95 standards
  • Typical applications include plastering, cement, sanding, and wood dust
  • Prevents against risky infectious diseases, according to national recommendations
  • Typical use time: 8 hours
Specifications

Specifications

EN 14683 Type: IIR
240 x 90 mm (± 3 mm)
N95
Polypropylene
Non-sterile
Made in France
Yes
Category 3 Personal Protective Equipment
Dust Mask
Transversal Elastic Ties
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit