Learn More
Cell Biolabs IncSupplier Diversity Partner HORAC ACTIVITY ASSAY TRL SIZE  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner STA346T
The OxiSelect HORAC Activity Assay (Trial Size) is a powerful tool to measure the hydroxyl radical antioxidant capacity of biomolecules. Results in less than 2 hours on a fluorescence plate reader. 48 wells/kit.
Catalog No. 50-673-542