Learn More
HORIBA ABX Pentra™ 400 Clinical Chemistry Analyzer Controls
Supplier: HORIBA 1220001786
This item is not returnable.
View return policy
Specifications
CE | |
CK Control | |
4 x 3 mL |
ABX Pentra™ C400 | |
Serum, Plasma, Whole Blood or Hemolysate (HbA1c), and Urine (Microalbumin) | |
Lyophilized |
Safety and Handling
ShelfLife : 5 Years