Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HORIBA ISE Reagents for Pentra™ 400 Clinical Chemistry Analyzer
For use with Pentra 400 Clinical Chemistry Analyzer.
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity & Availability | ||||
23-600-481
|
HORIBA
1220001717 |
280 mL | Standard 1 |
Each for $142.22
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-600-482
|
HORIBA
1220001718 |
100 mL | Standard 2 |
Each for $130.94
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-600-483
|
HORIBA
1220001719 |
100 mL | Reference |
Each for $105.03
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- For use with Pentra 400 Clinical Chemistry Analyzer
- Formulation: RTU
- Container: Bottle
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title