HORIBA ABX Pentra™ 60 C + Hematology Analyzer
Five-part leukocyte differential analysis through a combination of cytochemistry, light absorbency technology, and focused-flow volumetric determination with selectable CBC or CBC plus five-part differential analysis
$59623.02
  • Products 1
  • Description & Specifications
Products
Catalog Number Mfr. No. Description Price Quantity Availability  
23-450-042
ABX Diagnostics
P60CP001NUA1
Closed tube; 26 parameters
Each for $59,623.02
Check Availability
Add to Cart
Descriptions & Specifications
Related Products