HORIBA ABX™ Reagents for ABX Pentra C200 Available on GSA/VA Contract for Federal Government customers only.

For ABX Pentra C200

Manufacturer: ABX Diagnostics, Inc. 1220001674

 View more versions of this product

Catalog No. 23-600-720

  Check Availability
Add to Cart

    Specifications

    Product Type Urine control L/H
    For Use With (Equipment) ABX Pentra™ C400
    Level High, Low
    Format RTU
    For Use With (Application) Serum, Plasma, Whole Blood or Hemolysate (HbA1c), and Urine (Microalbumin)
    Quantity 1 x 10mL + 1 x 10mL,1 x 10mL + 1 x 10mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit