Learn More
HORIBA ABX Reagents for ABX Pentra C400
FDA cleared Liquid Chemistry
$68.80 - $889.56
Specifications
For Use With (Application) | Serum, Plasma, Whole Blood or Hemolysate (HbA1c), and Urine (Microalbumin) |
---|
Catalog Number | Mfr. No. | Type | Form | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Form | Quantity | Price | Quantity & Availability | |||
23-600-700
|
HORIBA
1220001871 |
General Chemistry Reagent | Liquid | 125 Tests |
Each for $85.46
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-601-500
|
HORIBA
1220001648 |
CO2 Calibrator | Ready-to-Use (RTU) | 3 x 3mL |
Each for $68.80
|
|
|||
23-601-501
|
HORIBA
5301101890 |
CO2 Control | Ready-to-Use (RTU) | 4 x 3mL + 4 x 3mL |
Each for $889.56
|
|
|||
23-601-000
|
HORIBA
1220001901 |
Reference Reagent | Ready-to-Use (RTU) | 280 mL |
Each for $269.83
|
|
|||
Description
- With extended linearity and increased stability
- Standardized specific proteins
- Purified and concentrated antiserums
Compliance
Complies with international recommendations (IFCC)
Specifications
Serum, Plasma, Whole Blood or Hemolysate (HbA1c), and Urine (Microalbumin) |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.