HORIBA General Chemistry Reagents for ABX Pentra™ 400 Clinical Chemistry Analyzers Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: ABX Diagnostics, Inc. 1220001635

 View more versions of this product

Catalog No. 23-600-426

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Product Type Direct Bilirubin CP substrate
    For Use With (Equipment) ABX Pentra™ 400 clinical chemistry analyzer
    Formulation DCA
    Quantity 100 Tests

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit