Learn More
PicoStudio Horizontal Turbine Products for Science Education
The Horizontal Turbine teaches how wind turbines are designed and built, and how they generate energy.
$34.99 - $109.99
Specifications
Product Type STEM Kit
View More Specs
Alert:
Small Parts - Not recommended for children under 3 years old.
Products 3
Catalog Number Mfr. No. Description Includes Price Quantity & Availability  
Catalog Number Mfr. No. Description Includes Price Quantity & Availability  
S4131907 Products for Science Education
View Documents
PicoStudio
PS30011
Level 1 1 Base, 1 Short Tower 5.5 in. (139.7 mm), 1 Nacelle Base, 1 Nacelle Front, 1 Nose Cone, 2 Blade Hubs, 6 Blade Adapters, 6 Blades 6 in. (152 mm), 1 DC Shaft, 1 DC Generator Pack (1 DC Motor, 1 DC Motor Stand), 1 Alligator Clip Set (Black and Red), 1 Screw Pack (1 LED, 1 Piezo Buzzer, 6 Screws, 6 Nuts, 1 Resistor 30 Ohm, 1 Motor Clip)
Each for $34.99
Add to cart
S4131908 Products for Science Education
View Documents
PicoStudio
PS30012
Level 2 Horizontal Level 1 Parts, 1 Long Tower, 1 Weighted Base Insert, 1 AC Generator Pack (1 Coil Housing, 1 AC Shaft, 1 Magnet Clip Set, 2 O-Rings, 1 4-Count Coil Pack, 1 4-Count Magnet Pack), 1 Rectification Lab (1 Solderless Breadboard, 1 Piezo Buzzer, 8 Male End Wires, 2 Capacitors, 4 Diodes)
Each for $79.99
Add to cart
S4131909 Products for Science Education
View Documents
PicoStudio
PS30013
Level 3 Horizontal Level 2 Parts, 1 AC Generator Pack, 1 Rectification Lab, 1 Long Tower, 1 Weighted Base Insert, 1 Circuit 101 Lab (3 Resistors 10, 30, 100 Ohm, 2 AA Battery Holder, 1 Light Bulb 1.5V, 8 Male End Wires, 1 Light Bulb Stand, 1 Alligator Clips Set (Red and Black), 1 Multimeter), 1 Solar Panel 7.2V 200mA
Each for $109.99
Add to cart
Description
Description
  • Explore the power of wind energy with our Horizontal Wind Turbine!
  • Learn how wind turbines are designed and built, and how they generate energy.
Specifications
Specifications
STEM Kit