Learn More
Aviva Systems Biology Corp HORMAD2 Antibody - middle region (ARP53008_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53008_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HORMAD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HORMAD2. Alternate names: MGC26710ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-60894