Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hormone-sensitiveLipase/HSL Rabbit anti-Human, Clone: SN06-39, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP267655
Description
Hormone-sensitive Lipase/HSL Monoclonal specifically detects Hormone-sensitive Lipase/HSL in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Hormone-sensitiveLipase/HSL | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000-1:5000, Immunohistochemistry, Immunohistochemistry-Paraffin | |
hormone-sensitive lipase, hormone-sensitive lipase testicular isoform, HSLEC 3.1.1.79, LHS, lipase, hormone-sensitive | |
Rabbit | |
Protein A purified | |
Cancer, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism, Lipid Droplets, Phospho Specific, Signal Transduction | |
3991 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
SN06-39 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
LIPE | |
Synthetic phospho-peptide corresponding to residues surrounding Ser853 of Human Hormone-sensitive Lipase/HSL aa 821-870/ 1,076. (SwissProt: Q05469 Human; SwissProt: P54310 Mouse; SwissProt: P15304 Rat) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction