Learn More
Crystal Chem IncSupplier Diversity Partner Horse Estrone-3-Sulfate ELISA Kit, 96-wells DFS Item
Manufacturer:  Crystal Chem IncSupplier Diversity Partner 80567
The Horse Estrone-3-Sulfate ELISA is used to quantify levels of horse estrone-3-sulfate in serum using only a 20 µL sample volume. Highlights: Kit uses sample size of only 25 µL. Wide dynamic range of 5 - 1000 ng/mL. High sensitivity of 0.14 ng/mL)
Catalog No. 50-194-7735