Learn More
Cole-Parmer CPC (Colder) PMCD220412 Quick-Disconnect Hose Barb Insert; Valved, 1/4" Tube ID, 1/8" Flow Size, PP  DFS Item
Manufacturer:  Cole-Parmer 0636445
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting—fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.
Catalog No. 50-628-255