Learn More
Nordon Hoshizaki™ Ice Machine and Cuber
Nordon Hoshizaki Ice machine and cuber features a Front in, front out airflow system that eliminates need for side and rear clearance.
Supplier: Nordon KM151BWH
Description
- Compact undercounter top hat Ice maker
- Features a Front in, front out airflow system that eliminates need for side and rear clearance
- Perfect for fitting into tight spaces. Drain pump not included
- Perfect addition to any hotel suite, office breakroom, or residential kitchen
Specifications
58.96 kg | |
25 gal. | |
130 lb. | |
130 lb. | |
45° to 100°F | |
99.1cm | |
39 in. |
3 Years | |
AHRI, ETL, Energy Star™ and NSF | |
6.8A | |
7.22° to 37.77°C | |
23.75 in. | |
115V |