Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nordon Hoshizaki™ Ice Machine and Dispenser

Nordon Hoshizaki Ice machine and dispenser features the CycleSaver? design and includes the EverCheck? alert system making unit simple to maintain

Manufacturer:  Nordon KM260BWH

 View more versions of this product

Catalog No. 11-675-104


  Check Availability
Add to cart

Specifications

Description

  • Undercounter water-cooled, self contained
  • Built in storage bin ice maker stores up to 80 lbs of ice
  • Ideal for small bars, hotel lobbies, and office break rooms
  • Features the CycleSaver™ design and includes the EverCheck™ alert system making unit simple to maintain
  • Requires a Single phase, 115v/60/1 electrical connection
Description & Specifications

Specifications

45° to 100°F
7.5A
AHRI, ETL, NSF and UL
99.1cm
3 Years
30 in.
121.56kg
7.22° to 37.77°C
268 lb.
39 in.
115V
26.3 gal.
268 lb.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit