Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Host Cell Factor 1/HCFC1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NB10068209
Description
Host Cell Factor 1/HCFC1 Polyclonal specifically detects Host Cell Factor 1/HCFC1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Knockdown Validated.Specifications
Host Cell Factor 1/HCFC1 | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
CFFC1 factor, HCF, HCF1HFC1, HCF-1VP16 accessory protein, host cell factor C1 (VP16-accessory protein), MGC70925, VCAFhost cell factor 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
3054 | |
Human, Mouse | |
IgG |
Western Blot, Immunoprecipitation, KnockDown | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate, KnockDown Validated | |
P51610 | |
HCFC1 | |
The immunogen recognized by this antibody maps to a region between residue 1700 and 1750 of human host cell factor C1 (VP16-accessory protein) using the numbering given in entry NP_005325.2 (GeneID 3054). | |
0.1 mL | |
Primary | |
Human. May react with Mouse. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction