Thermo Scientific™ Hot Gas Bypass Temperature Controllers
Combine these hot gas bypass temperature controllers with Thermo Scientific™ CryoPlus™ LN2 cryopreservation storage systems for increased utility.
Manufacturer: Thermo Scientific™ 195023
 View more versions of this product
Catalog No. 13-262-793
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) CryoPlus 1,2,3,4
    Type Factory-Installed,Temperature Controller
    Description Factory installed, 220V
    For Use With CryoPlus 1,2,3,4
    Combine Thermo Scientific™ Hot Gas Bypass Temperature Controllers with Thermo Scientific™ CryoPlus™ LN2 cryopreservation storage systems for increased utility.
    Features:
    • 120V or 220V
    • Factory or field installed
    Ordering Information:
    Field installed option must be installed by a qualified professional.
  • Description & Specifications
Related Products
Product Content Feedback