Learn More
Chemglass Life Sciences Digital Magnetic Hot Plate with Timer Bundles, with Tri-Block, Vials, COMBO 1 - Hotplate and Tri-block
SDP
Supplier:  Chemglass Life Sciences CG1997V210
Encompass_Preferred
Digital Magnetic Hot Plate with Timer Bundles, with Tri-Block, Vials, COMBO 1 - Hotplate and Tri-block.Reaction TRI-BLOCK is designed to fit directly on 135mm diameter circular top hot plate stirrers. Versatile design of the tri-block fits the most common vial sizes on one hot plate: 11ea- 1 Dram vials, 11ea- 2 Dram vials and 7ea– 20mL scintillation vials. Tri-block is hard coat anodized black aluminum. A drilled thermowell is provided for insertion of the PT-100 Temperature Sensor. Custom configurations are available. Please call for pricing.Designed with Advanced Features• Integral timer - settable from 1 minute to 99 hours, 59 minutes• Service life meter - track service interval or service lifetime• Bright white LED display for high-contrast easy reading• 135mm diameter, ceramic-coated, aluminum alloy plateCOMBO 1 Includes 1 each:• CG-1997-V-10• CG-1991-T-10• Trial Pack of CG-4912 Pressure Relief Reaction Vials
Catalog No. 50-135-4669
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.